| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016307.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016307.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016307.hg.1 |
TC11001286.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016308.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016308.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016308.hg.1 |
TC11001286.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009817.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009817.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009817.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009817.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174160.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174160.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174160.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174160.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174160.hg.1 |
TC1100009817.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009817.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009817.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009817.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009817.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009817.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174159.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174159.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174159.hg.1 |
ENST00000314222.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174159.hg.1 |
NM_003311.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174159.hg.1 |
TC1100009817.hg.1 |
Clariom D TC |