Search
Search

FAM209B| Gene Symbol: FAM209B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013646.3 | NP_001013668.2 | 2 | 446 | 122 | |
| XM_017027849.1 | XP_016883338.1 | 4 | 962 | 122 | ||
| GenBank mRNA | BC105792.1 | - | 3 | 411 | 6618487 | 122 |
| DB452055.1 | - | 2 | 407 | 122 | ||
| DB517586.1 | - | 2 | 211 | 122 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17202242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880561 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11230702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11230693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11230701 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880539 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11230705 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880550 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17202263 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880560 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880567 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11230682 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880547 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880555 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11230686 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11230678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880516 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11230706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880512 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17202267 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31880549 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11230681 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 567716 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 271306 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3404014 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0992 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P305533 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0993 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 32956 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 9960 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03538389 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P41363573 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3404014 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3404014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17202250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880514 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230685 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17202242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230685 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17202250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17202242 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11230680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880514 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31880515 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P305533 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P03538389 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230679 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230684 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230674 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230676 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230685 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230686 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230680 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230678 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230683 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230673 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11230682 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03538389 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P305533 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 104765 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 74400 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21168248 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03538389 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230704 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230702 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17202268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230695 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230697 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230685 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230689 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230671 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230678 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230693 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17202238 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230679 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230676 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230700 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17202229 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230687 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230674 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230706 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230692 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11230682 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03538389 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001013646_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8063532 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8063533 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8063531 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 232513_PM_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009946.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009946.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009946.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009946.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009946.hg.1 | NM_001013646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009946.hg.1 | NM_001013646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009946.hg.1 | NM_001013646.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009946.hg.1 | NM_001013646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | ENST00000371328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009945.hg.1 | NM_001012971.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170248.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170248.hg.1 | NM_001013646.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170248.hg.1 | TC2000009946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000170248.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170248.hg.1 | NM_001013646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009946.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009946.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009946.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009946.hg.1 | ENST00000371325.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009946.hg.1 | NM_001013646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009946.hg.1 | NM_001013646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009946.hg.1 | NM_001013646.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009946.hg.1 | NM_001013646.3 | Refseq |
| Human Genome U133 Plus 2.0 Array | 232513_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 233086_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 233086_at | - | - |
| Human Genome U133 Plus 2.0 Array | 232513_PM_x_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001013646_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232513_x_at6 | - | - |
| Human Genome U95 Set | 76219_PM_at | - | - |
| Human Genome U95 Set | 76219_at | - | - |
| Human Genome U133 Set | 233086_PM_at | - | - |
| Human Genome U133 Set | 232513_PM_x_at | - | - |
| Human Genome U133 Set | 233086_at | - | - |
| Human Genome U133 Set | 232513_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018477 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018477 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1714667 | BC014951.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1714667 | CR593795.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1714667 | NM_001013646.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2148497 | BC105792.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2148497 | CR593795.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2148497 | NM_001013646.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1714667 | BC014951.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1714667 | CR593795.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1714667 | NM_001013646.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1714667 | BC014951.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1714667 | CR593795.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1714667 | NM_001013646.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2148497 | BC105792.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2148497 | CR593795.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2148497 | NM_001013646.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2148497 | BC105792.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2148497 | CR593795.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2148497 | NM_001013646.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2148497 | BC105792.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2148497 | CR593795.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2148497 | NM_001013646.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_1548-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1714667 | BC014951.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1714667 | CR593795.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1714667 | NM_001013646.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1714667 | BC014951.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1714667 | CR593795.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1714667 | NM_001013646.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1450110 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_1548 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2148497 | BC105792.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2148497 | CR593795.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2148497 | NM_001013646.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2148497 | BC105792.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2148497 | CR593795.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2148497 | NM_001013646.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1714667 | BC014951.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1714667 | CR593795.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1714667 | NM_001013646.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2148497 | BC105792.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2148497 | CR593795.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2148497 | NM_001013646.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1714667 | BC014951.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1714667 | CR593795.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1714667 | NM_001013646.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1714667 | BC014951.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1714667 | CR593795.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1714667 | NM_001013646.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2148497 | BC105792.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2148497 | CR593795.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2148497 | NM_001013646.2 | human_transcriptome |