Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231538_s1
          See other MIR200A GE Assays ›
          Gene Symbol:
          MIR200A
          Gene Name:
          microRNA 200a
          Gene Aliases:
          MIRN200A, mir-200a
          Chromosome Location:
          Chr.1: 1167863 - 1167952 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231538_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR200A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029834.1 1 48 64
          GenBank mRNA LM608664.1 - 1 48 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0100018153.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000028.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          gene silencing by miRNA
          positive regulation of neuron apoptotic process
          positive regulation of blood vessel endothelial cell migration
          cellular response to beta-amyloid
          mRNA binding involved in posttranscriptional gene silencing

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001127 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001128 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001127 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001128 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 7896861 - -
          Affymetrix Human Transcriptome Array 2.0 TC0100018153.hg.1 ENST00000384875.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018153.hg.1 NR_029834.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01000306.hg.1 ENST00000384875.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01000306.hg.1 NR_029834.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01000306.hg.1 TC01000028.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100236670.hg.1 ENST00000384875.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100236670.hg.1 NR_029834.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100236670.hg.1 ENST00000384875.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100236670.hg.1 NR_029834.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100236670.hg.1 TC0100018153.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100018153.hg.1 ENST00000384875.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018153.hg.1 NR_029834.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308803 NR_029834.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308803 NR_029834.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308803 NR_029834.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308803 NR_029834.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline