Search
Search

MIR3125| Gene Symbol: MIR3125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036072.1 | 1 | 42 | 67 | ||
| GenBank mRNA | LM610972.1 | - | 1 | 42 | 67 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tribbles pseudokinase 2 | TRIB2 |
C5FW, GS3955, TRB2 |
Sense | Chr. 2 - 12716936 - 12742734 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100506474 | LOC100506474 | 229.3kb | Anti-sense | Chr. 2 - 12966782 - 13007013 on NCBI Build GRCh38 |
Set Membership: |
|