Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010010.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010010.hg.1 |
TC16000704.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
ENST00000562298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
ENST00000562298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
ENST00000562298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
ENST00000562298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008898.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600166608.hg.1 |
ENST00000562298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600166608.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600166608.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600166608.hg.1 |
ENST00000562298.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600166608.hg.1 |
ENST00000562866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600166608.hg.1 |
NR_036480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600166608.hg.1 |
TC1600008898.hg.1 |
Clariom D TC |