Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04233790_s1
          See other LINC00028 GE Assays ›
          Gene Symbol:
          LINC00028
          Gene Name:
          long intergenic non-protein coding RNA 28
          Gene Aliases:
          C20orf93, NCRNA00028, dJ1093G12.6
          Chromosome Location:
          Chr.20: 31485778 - 31487574 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04233790_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00028
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024358.1 1 48 68
          GenBank mRNA BX280465.1 - 1 48 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000007074.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000189.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3411392 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4530 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4529 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 8613 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3411392 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3411392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130445 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130444 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130440 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777518 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777522 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130441 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777524 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130443 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130443 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130444 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777522 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130445 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130441 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777524 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31777518 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11130440 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17186388 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130461 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130463 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130458 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130395 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130465 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130407 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130390 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130393 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17186392 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130467 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17186386 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130401 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130397 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130405 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130441 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130438 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17186379 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130469 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130455 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130443 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130453 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130448 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130445 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130411 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130472 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130403 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11130450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8061540 - -
          Affymetrix Human Gene 1.0 ST Array 8061541 - -
          Affymetrix Human Gene 1.1 ST Array 8061539 - -
          Affymetrix HT HG-U133+ PM Array Plate 233812_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR20002261.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002261.hg.1 NR_024358.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002261.hg.1 TC20000189.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002259.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002259.hg.1 NR_024358.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002259.hg.1 TC20000189.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002262.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002262.hg.1 NR_024358.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002262.hg.1 TC20000189.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20002260.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20002260.hg.1 NR_024358.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20002260.hg.1 TC20000189.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000007074.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007074.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007074.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007074.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007074.hg.1 NR_024358.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007074.hg.1 NR_024358.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007074.hg.1 NR_024358.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007074.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148243.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148243.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148243.hg.1 TC2000007074.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148243.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148243.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148240.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148240.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148240.hg.1 TC2000007074.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148240.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148240.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007074.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007074.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007074.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007074.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007074.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007074.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007074.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay TC2000007074.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148238.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148238.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148238.hg.1 TC2000007074.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148238.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148238.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148241.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148241.hg.1 NR_024358.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000148241.hg.1 TC2000007074.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000148241.hg.1 ENST00000435497.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000148241.hg.1 NR_024358.1 Refseq
          Human Genome U133 Plus 2.0 Array 233812_at - -
          Human Genome U133 Plus 2.0 Array 233812_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233812_at2 - -
          Human Genome U133 Set 233812_at - -
          Human Genome U133 Set 233812_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018179 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A018179 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3248428 NR_024358.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3248428 AI675392.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3248428 BX280465.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248428 NR_024358.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248428 AI675392.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248428 BX280465.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248428 NR_024358.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248428 AI675392.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248428 BX280465.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248428 NR_024358.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248428 AI675392.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248428 BX280465.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline