Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273238_s1
          See other MIR148A GE Assays ›
          Gene Symbol:
          MIR148A
          Gene Name:
          microRNA 148a
          Gene Aliases:
          MIRN148, MIRN148A, hsa-mir-148, mir-148a
          Chromosome Location:
          Chr.7: 25949919 - 25949986 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273238_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 4,565.00
          Your Price
          Online offer:
          4565.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR148A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029597.1 1 28 66

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC646588 LOC646588 -279.2kb Sense Chr. 7 - 25593301 - 25790051 on NCBI Build GRCh38
          Downstream nuclear factor,
          erythroid 2 like 3
          NFE2L3 NRF3 202.2kb Anti-sense Chr. 7 - 26152227 - 26187137 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0700010531.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001213.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372011 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589201 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589195 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589212 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05371999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05371994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589225 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15919158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15919161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589199 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589209 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15919165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589201 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589209 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372011 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15919158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589195 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589199 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589212 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05371994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589225 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15919165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15919161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05371999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05372017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25589214 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8138668 - -
          Affymetrix Human Transcriptome Array 2.0 TC0700010531.hg.1 ENST00000362215.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010531.hg.1 NR_029597.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07018959.hg.1 TC07001213.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07018959.hg.1 ENST00000362215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07018959.hg.1 NR_029597.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010531.hg.1 ENST00000362215.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700010531.hg.1 NR_029597.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700174886.hg.1 ENST00000362215.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700174886.hg.1 NR_029597.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700174886.hg.1 ENST00000362215.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700174886.hg.1 NR_029597.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700174886.hg.1 TC0700010531.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308723 NR_029597.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308723 NR_029597.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308723 NR_029597.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308723 NR_029597.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline