Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274374_m1
          See other STEAP1B-AS1 GE Assays ›
          Gene Symbol:
          STEAP1B-AS1
          Gene Name:
          STEAP1B antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 22563337 - 22573998 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274374_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: STEAP1B-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038393.1 2 - 3 276 106
          GenBank mRNA AK123961.1 - 2 - 3 269 106
          DA428433.1 - 2 - 3 276 106
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA888584.1 - 2 - 3 269 Chr. 7 - 22563344 - 22572144 [+] on Build GRCh38 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream STEAP family member 1B STEAP1B -64.8kb Anti-sense Chr. 7 - 22419444 - 22500188 on NCBI Build GRCh38
          Downstream uncharacterized LOC401312 LOC401312 77.9kb Anti-sense Chr. 7 - 22649960 - 22665533 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700006885.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000135.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356972 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573803 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15917123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573773 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356946 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573811 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573748 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356967 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573766 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356985 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573812 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356965 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356973 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573784 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573770 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15917131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573761 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573809 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357027 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15917124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573810 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356989 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573827 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05357002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573826 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25573779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05356988 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P01672811 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01672811 - -
          Agilent Human 1B Microarray G4111A A_32_P178247 - -
          Agilent Human 1B Microarray G4111A A_32_P55480 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07002109.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07002109.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07002109.hg.1 TC07000135.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07002110.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07002110.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07002110.hg.1 TC07000135.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07002107.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07002107.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07002107.hg.1 TC07000135.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000439823.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000439823.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000439823.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 ENST00000439823.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07002108.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07002108.hg.1 NR_038393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07002108.hg.1 TC07000135.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700147613.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147613.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147613.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147613.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147613.hg.1 TC0700006885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700147603.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147603.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147603.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147603.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147603.hg.1 TC0700006885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700147615.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147615.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147615.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147615.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147615.hg.1 TC0700006885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700147614.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147614.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147614.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147614.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147614.hg.1 TC0700006885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700147608.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147608.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147608.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147608.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147608.hg.1 TC0700006885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700147611.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147611.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147611.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147611.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147611.hg.1 TC0700006885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000439823.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000439823.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000439823.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 ENST00000439823.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay TC0700006885.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147604.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147604.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147604.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147604.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147604.hg.1 TC0700006885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700147602.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147602.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147602.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147602.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147602.hg.1 TC0700006885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700147601.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147601.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147601.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147601.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147601.hg.1 TC0700006885.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700147612.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147612.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147612.hg.1 ENST00000435127.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147612.hg.1 NR_038393.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700147612.hg.1 TC0700006885.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A034042 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A034042 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline