| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012164.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012164.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012164.hg.1 |
NR_038854.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012164.hg.1 |
NR_038854.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026876.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026876.hg.1 |
NR_038854.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026876.hg.1 |
TC12002046.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026875.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026875.hg.1 |
NR_038854.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026875.hg.1 |
TC12002046.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200194884.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200194884.hg.1 |
NR_038854.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200194884.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200194884.hg.1 |
NR_038854.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200194884.hg.1 |
TC1200012164.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200194885.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200194885.hg.1 |
NR_038854.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200194885.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200194885.hg.1 |
NR_038854.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200194885.hg.1 |
TC1200012164.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200012164.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012164.hg.1 |
ENST00000500741.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012164.hg.1 |
NR_038854.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012164.hg.1 |
NR_038854.1 |
Refseq |