Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000591875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000591875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000591875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000591875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012245.hg.1 |
ENST00000315997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012245.hg.1 |
ENST00000350219.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012245.hg.1 |
ENST00000422873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012245.hg.1 |
ENST00000472572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012245.hg.1 |
ENST00000474471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012245.hg.1 |
ENST00000477550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012245.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012245.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012245.hg.1 |
TC16000841.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000591875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000591875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000591875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000591875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009257.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000315997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000472572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000474471.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000586629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
ENST00000592343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
NM_001098514.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
NM_152459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600171197.hg.1 |
TC1600009257.hg.1 |
Clariom D TC |