| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011783.hg.1 |
ENST00000399974.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011783.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011783.hg.1 |
TC16000815.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170128.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170128.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170128.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170128.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170128.hg.1 |
TC1600009192.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600170129.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170129.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170129.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170129.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170129.hg.1 |
TC1600009192.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009192.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170127.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170127.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170127.hg.1 |
ENST00000399974.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170127.hg.1 |
NM_001190476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170127.hg.1 |
TC1600009192.hg.1 |
Clariom D TC |