| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017091.hg.1 |
TC08001327.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017091.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010802.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173344.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173344.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173344.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173344.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173344.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173344.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173344.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173344.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173344.hg.1 |
TC0800010802.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800173345.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173345.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173345.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173345.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173345.hg.1 |
ENST00000613105.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173345.hg.1 |
ENST00000624510.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173345.hg.1 |
ENST00000625134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173345.hg.1 |
NM_001243237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173345.hg.1 |
TC0800010802.hg.1 |
Clariom D TC |