| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008303.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008303.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008303.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008303.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008303.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008303.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008303.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008303.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009013.hg.1 |
ENST00000441889.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009013.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009013.hg.1 |
TC17000687.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009011.hg.1 |
ENST00000441889.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009011.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009011.hg.1 |
TC17000687.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009012.hg.1 |
ENST00000441889.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009012.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009012.hg.1 |
TC17000687.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700161895.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700161895.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700161895.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700161895.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700161895.hg.1 |
TC1700008303.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700161893.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700161893.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700161893.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700161893.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700161893.hg.1 |
TC1700008303.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700161894.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700161894.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700161894.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700161894.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700161894.hg.1 |
TC1700008303.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700008303.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008303.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008303.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008303.hg.1 |
ENST00000623702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008303.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008303.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008303.hg.1 |
NM_001243552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008303.hg.1 |
NM_001243552.1 |
Refseq |