Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04285637_m1
          See other SALL1 GE Assays ›
          Gene Symbol:
          SALL1
          Gene Name:
          spalt like transcription factor 1
          Gene Aliases:
          HEL-S-89, HSAL1, Sal-1, TBS, ZNF794
          Chromosome Location:
          Chr.16: 51135975 - 51152386 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04285637_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SALL1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127892.1 NP_001121364.1 2 - 3 3672 79
          NM_002968.2 NP_002959.2 2 - 3 3562 79
          XM_006721241.3 XP_006721304.1 3 - 4 3637 79
          XM_011523254.2 XP_011521556.1 3 - 4 3663 79
          GenBank mRNA AK295103.1 - 3 - 4 3452 79
          EU668343.1 - 2 - 3 3531 79
          Y18265.1 - 2 - 3 3531 79

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SALL1

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ureteric bud development
          branching involved in ureteric bud morphogenesis
          kidney development
          ventricular septum development
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          transcription, DNA-templated
          heart development
          gonad development
          histone deacetylation
          olfactory nerve development
          olfactory bulb interneuron differentiation
          pituitary gland development
          neurogenesis
          positive regulation of Wnt signaling pathway
          adrenal gland development
          inductive cell-cell signaling
          somatic stem cell population maintenance
          outer ear morphogenesis
          embryonic digit morphogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic digestive tract development
          limb development
          olfactory bulb mitral cell layer development
          kidney epithelium development
          ureteric bud invasion
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          histone deacetylase activity
          protein binding
          beta-catenin binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000251020.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000440970.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000562674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000566102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 ENST00000570206.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_001127892.1 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010291.hg.1 NM_002968.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline