Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04331211_s1
          See other SNORD1B GE Assays ›
          Gene Symbol:
          SNORD1B
          Gene Name:
          small nucleolar RNA, C/D box 1B
          Gene Aliases:
          R38B, snR38B
          Chromosome Location:
          Chr.17: 76561108 - 76561193 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04331211_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORD1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_004396.1 1 48 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA host gene 16 SNHG16 ncRAN Sense Chr. 17 - 76557764 - 76565348 on NCBI Build GRCh38
          Downstream small nucleolar RNA,
          C/D box 1A
          SNORD1A R38A,
          snR38A
          440.0 Sense Chr. 17 - 76561633 - 76561706 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008955.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000876.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10471395 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31055797 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10471391 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31055793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10471397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10471394 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10471397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31055797 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10471395 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10471391 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31055793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10471394 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471397 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471383 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471388 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471372 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471394 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471370 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471381 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471378 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17018136 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471386 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471361 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471376 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471367 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471374 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471392 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471365 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471391 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471385 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471363 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471382 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471379 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471395 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471364 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10471380 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17018132 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_004396_at - -
          Affymetrix Human Gene 1.0 ST Array 8010081 - -
          Affymetrix Human Gene 1.1 ST Array 8010080 - -
          Affymetrix Human Transcriptome Array 2.0 TC1700008955.hg.1 ENST00000363091.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008955.hg.1 NR_004396.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17011386.hg.1 ENST00000363091.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17011386.hg.1 NR_004396.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17011386.hg.1 TC17000876.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1700008955.hg.1 ENST00000363091.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008955.hg.1 NR_004396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700167506.hg.1 ENST00000363091.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167506.hg.1 NR_004396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700167506.hg.1 ENST00000363091.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167506.hg.1 NR_004396.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700167506.hg.1 TC1700008955.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3245554 NR_004396.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3245554 NR_004396.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245554 NR_004396.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3245554 NR_004396.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline