Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04332240_s1
          See other RPS16P5 GE Assays ›
          Gene Symbol:
          RPS16P5
          Gene Name:
          ribosomal protein S16 pseudogene 5
          Gene Aliases:
          RPS16_3_702
          Chromosome Location:
          Chr.6: 53334460 - 53337617 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04332240_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RPS16P5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046241.1 1 3120 96
          GenBank mRNA AK097937.2 - 1 3120 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600012073.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3209254 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 8906 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3209254 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1566079_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0600012073.hg.1 ENST00000406501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012073.hg.1 NR_046241.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012073.hg.1 NR_046241.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012073.hg.1 NR_046241.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183123.hg.1 NR_046241.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183123.hg.1 NR_046241.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183123.hg.1 TC0600012073.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600183122.hg.1 ENST00000406501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183122.hg.1 NR_046241.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183122.hg.1 ENST00000406501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183122.hg.1 NR_046241.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183122.hg.1 TC0600012073.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600183121.hg.1 NR_046241.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183121.hg.1 NR_046241.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183121.hg.1 TC0600012073.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600012073.hg.1 ENST00000406501.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012073.hg.1 NR_046241.1 Refseq
          Affymetrix Human Clariom D Assay TC0600012073.hg.1 NR_046241.1 Refseq
          Affymetrix Human Clariom D Assay TC0600012073.hg.1 NR_046241.1 Refseq
          Human Genome U133 Plus 2.0 Array 1566079_PM_at - -
          Human Genome U133 Plus 2.0 Array 1566079_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566079_at5 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline