| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
231551_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013266.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035997.hg.1 |
TC01002334.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035997.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035996.hg.1 |
TC01002334.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035996.hg.1 |
ENST00000440767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035996.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035994.hg.1 |
TC01002334.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035994.hg.1 |
ENST00000440767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035994.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198338.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198338.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198338.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198338.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198338.hg.1 |
TC0100013266.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013266.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198334.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198334.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198334.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198334.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198334.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198334.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198334.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198334.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198334.hg.1 |
TC0100013266.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100198331.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198331.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198331.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198331.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198331.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198331.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198331.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198331.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198331.hg.1 |
TC0100013266.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100198332.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198332.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198332.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198332.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198332.hg.1 |
ENST00000440767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198332.hg.1 |
ENST00000566855.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198332.hg.1 |
ENST00000622840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198332.hg.1 |
NM_001242521.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198332.hg.1 |
TC0100013266.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
231551_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
231551_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
231551_at1 |
- |
- |
| Human Genome U95 Set |
76645_PM_at |
- |
- |
| Human Genome U95 Set |
87949_at |
- |
- |
| Human Genome U95 Set |
87949_PM_at |
- |
- |
| Human Genome U95 Set |
76645_at |
- |
- |
| Human Genome U133 Set |
231551_PM_at |
- |
- |
| Human Genome U133 Set |
231551_at |
- |
- |