Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04370938_g1
          See other ANAPC15 GE Assays ›
          Gene Symbol:
          ANAPC15
          Gene Name:
          anaphase promoting complex subunit 15
          Gene Aliases:
          APC15, C11orf51, HSPC020
          Chromosome Location:
          Chr.11: 72106372 - 72112780 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04370938_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANAPC15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001278485.2 NP_001265414.1 3 - 4 267 76
          NM_001278486.2 NP_001265415.1 3 - 4 262 76
          NM_001278487.2 NP_001265416.1 3 - 4 262 76
          NM_001278488.2 NP_001265417.1 3 - 4 263 76
          NM_001278489.2 NP_001265418.1 3 - 4 237 76
          NM_001278490.2 NP_001265419.1 2 - 3 182 76
          NM_001278491.1 NP_001265420.1 3 - 4 246 76
          NM_001278492.2 NP_001265421.1 3 - 4 236 76
          NM_001278493.2 NP_001265422.1 3 - 4 281 76
          NM_001278494.2 NP_001265423.1 3 - 4 533 76
          NM_001330321.2 NP_001317250.1 3 - 4 267 76
          NM_001393427.1 NP_001380356.1 3 - 4 262 76
          NM_001393428.1 NP_001380357.1 3 - 4 237 76
          NM_001393429.1 NP_001380358.1 2 - 3 182 76
          NM_001393430.1 NP_001380359.1 2 - 3 177 76
          NM_001393431.1 NP_001380360.1 3 - 4 533 76
          NM_001393432.1 NP_001380361.1 2 - 3 182 76
          NM_001393433.1 NP_001380362.1 3 - 4 267 76
          NM_001393434.1 NP_001380363.1 3 - 4 241 76
          NM_001393435.1 NP_001380364.1 3 - 4 251 76
          NM_001393436.1 NP_001380365.1 3 - 4 533 76
          NM_001393437.1 NP_001380366.1 3 - 4 268 76
          NM_001393438.1 NP_001380367.1 3 - 4 237 76
          NM_001393439.1 NP_001380368.1 2 - 3 182 76
          NM_001393440.1 NP_001380369.1 3 - 4 246 76
          NM_001393441.1 NP_001380370.1 3 - 4 528 76
          NM_001393442.1 NP_001380371.1 3 - 4 533 76
          NM_001393443.1 NP_001380372.1 3 - 4 267 76
          NM_001393444.1 NP_001380373.1 3 - 4 533 76
          NM_001393445.1 NP_001380374.1 3 - 4 528 76
          NM_001393446.1 NP_001380375.1 3 - 4 238 76
          NM_001393447.1 NP_001380376.1 3 - 4 268 76
          NM_001393448.1 NP_001380377.1 2 - 3 177 76
          NM_001393449.1 NP_001380378.1 3 - 4 241 76
          NM_001393450.1 NP_001380379.1 3 - 4 216 76
          NM_001393451.1 NP_001380380.1 3 - 4 528 76
          NM_001393452.1 NP_001380381.1 3 - 4 246 76
          NM_001393453.1 NP_001380382.1 3 - 4 251 76
          NM_001393454.1 NP_001380383.1 3 - 4 276 76
          NM_001393455.1 NP_001380384.1 2 - 3 448 76
          NM_001393456.1 NP_001380385.1 3 - 4 241 76
          NM_001393457.1 NP_001380386.1 3 - 4 503 76
          NM_001393458.1 NP_001380387.1 2 - 3 443 76
          NM_014042.3 NP_054761.1 3 - 4 267 76
          NR_171687.1 3 - 4 262 76
          XM_047426723.1 XP_047282679.1 3 - 4 216 76
          GenBank mRNA AF077206.1 - 3 - 4 349 76
          AK222622.1 - 3 - 4 335 76
          AL080071.1 - 3 - 4 352 76
          BC005156.2 - 3 - 4 341 76
          BC005393.1 - 3 - 4 342 76
          BE616852.1 - 3 - 4 247 76
          BG752082.1 - 3 - 4 327 76
          BG764016.1 - 3 - 4 330 76
          BU532056.1 - 3 - 4 263 76
          BU856100.1 - 3 - 4 292 76
          BU930156.1 - 2 - 3 176 76
          CD049337.1 - 3 - 4 555 76
          CN306434.1 - 3 - 4 318 76
          HY080534.1 - 3 - 4 245 76
          HY166245.1 - 3 - 4 343 76

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ANAPC15

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of mitotic cell cycle
          anaphase-promoting complex-dependent catabolic process
          cell division
          regulation of meiotic cell cycle
          protein K48-linked ubiquitination
          protein K11-linked ubiquitination
          regulation of mitotic cell cycle spindle assembly checkpoint
          protein branched polyubiquitination
          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline