Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04398572_m1
          See other TPM1 GE Assays ›
          Gene Symbol:
          TPM1
          Gene Name:
          tropomyosin 1
          Gene Aliases:
          C15orf13, CMD1Y, CMH3, HEL-S-265, HTM-alpha, LVNC9, TMSA
          Chromosome Location:
          Chr.15: 63042747 - 63071915 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04398572_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TPM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000366.6 NP_000357.3 4 - 5 576 59
          NM_001018004.2 NP_001018004.1 4 - 5 576 59
          NM_001018005.2 NP_001018005.1 4 - 5 576 59
          NM_001018006.2 NP_001018006.1 4 - 5 576 59
          NM_001018007.2 NP_001018007.1 4 - 5 576 59
          NM_001018008.2 NP_001018008.1 3 - 4 472 59
          NM_001018020.2 NP_001018020.1 4 - 5 576 59
          NM_001301244.2 NP_001288173.1 4 - 5 576 59
          NM_001301289.2 NP_001288218.1 3 - 4 472 59
          NM_001330344.2 NP_001317273.1 3 - 4 472 59
          NM_001330346.2 NP_001317275.1 3 - 4 472 59
          NM_001330351.2 NP_001317280.1 3 - 4 472 59
          NM_001365776.1 NP_001352705.1 4 - 5 576 59
          NM_001365777.1 NP_001352706.1 4 - 5 576 59
          NM_001365778.1 NP_001352707.1 5 - 6 702 59
          NM_001365779.1 NP_001352708.1 4 - 5 576 59
          NM_001365780.1 NP_001352709.1 3 - 4 472 59
          NM_001365781.2 NP_001352710.1 3 - 4 472 59
          NM_001365782.1 NP_001352711.1 3 - 4 472 59
          NM_001407322.1 NP_001394251.1 5 - 6 702 59
          NM_001407323.1 NP_001394252.1 5 - 6 702 59
          NM_001407324.1 NP_001394253.1 5 - 6 702 59
          NM_001407325.1 NP_001394254.1 4 - 5 576 59
          NM_001407326.1 NP_001394255.1 4 - 5 576 59
          NM_001407327.1 NP_001394256.1 4 - 5 576 59
          NM_001407328.1 NP_001394257.1 4 - 5 576 59
          NM_001407329.1 NP_001394258.1 4 - 5 576 59
          NM_001407330.1 NP_001394259.1 4 - 5 576 59
          NM_001407331.1 NP_001394260.1 4 - 5 576 59
          NM_001407332.1 NP_001394261.1 4 - 5 576 59
          NM_001407333.1 NP_001394262.1 4 - 5 576 59
          NM_001407334.1 NP_001394263.1 4 - 5 576 59
          NM_001407335.1 NP_001394264.1 4 - 5 576 59
          NM_001407336.1 NP_001394265.1 4 - 5 576 59
          NM_001407337.1 NP_001394266.1 4 - 5 576 59
          NM_001407338.1 NP_001394267.1 4 - 5 576 59
          NM_001407340.1 NP_001394269.1 3 - 4 472 59
          NM_001407341.1 NP_001394270.1 3 - 4 472 59
          NM_001407342.1 NP_001394271.1 3 - 4 472 59
          NM_001407344.1 NP_001394273.1 3 - 4 472 59
          NR_176337.1 4 - 5 576 59
          NR_176338.1 4 - 5 576 59
          NR_176339.1 5 - 6 671 59
          NR_176340.1 5 - 6 671 59
          NR_176341.1 4 - 5 576 59
          NR_176342.1 4 - 5 576 59
          NR_176343.1 4 - 5 576 59
          NR_176344.1 5 - 6 702 59
          NR_176345.1 4 - 5 576 59
          NR_176346.1 4 - 5 576 59
          NR_176347.1 4 - 5 576 59
          NR_176348.1 3 - 4 412 59
          NR_176349.1 3 - 4 412 59
          NR_176350.1 3 - 4 472 59
          NR_176351.1 3 - 4 472 59
          NR_176352.1 3 - 4 472 59
          NR_176353.1 3 - 4 472 59
          XM_047433001.1 XP_047288957.1 4 - 5 576 59
          GenBank mRNA AB209041.1 - 3 - 4 561 59
          AF474156.1 - 3 - 4 286 59
          AK057989.1 - 6 - 7 470 59
          AK092051.1 - 3 - 4 1401 59
          AK092573.1 - 2 - 3 1506 59
          AK131384.1 - 5 - 6 698 59
          AK298625.1 - 3 - 4 400 59
          AK299387.1 - 4 - 5 573 59
          AK301319.1 - 3 - 4 420 59
          AK309170.1 - 4 - 5 893 59
          AL050179.1 - 4 - 5 565 59
          AL558518.3 - 3 - 4 489 59
          BC007433.2 - 4 - 5 634 59
          BC050473.1 - 3 - 4 511 59
          BC053545.1 - 3 - 4 522 59
          BX458841.2 - 4 - 5 553 59
          DQ424903.1 - 3 - 4 385 59
          GQ472198.1 - 4 - 5 634 59
          KU178366.1 - 4 - 5 493 59
          KU178367.1 - 4 - 5 493 59
          M19714.1 - 4 - 5 435 59
          M19715.1 - 3 - 4 330 59
          X12369.1 - 3 - 4 323 59

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TPM1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of heart rate by epinephrine
          regulation of muscle contraction
          cytoskeleton organization
          actin filament organization
          regulation of heart contraction
          regulation of cell shape
          muscle filament sliding
          negative regulation of cell migration
          ruffle organization
          cellular response to reactive oxygen species
          wound healing
          sarcomere organization
          positive regulation of cell adhesion
          positive regulation of stress fiber assembly
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle contraction
          negative regulation of vascular smooth muscle cell proliferation
          negative regulation of vascular associated smooth muscle cell migration
          actin binding
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal protein binding
          structural constituent of muscle
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          actin filament binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline