Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04398857_g1
          See other MEMO1 GE Assays ›
          Gene Symbol:
          MEMO1 MEMO1P1
          Gene Name:
          mediator of cell motility 1
          MEMO1 pseudogene 1
          Gene Aliases:
          C2orf4, CGI-27, MEMO, NS5ATP7
          C21orf19, C2orf4P, MEMO1P, rcC2orf4
          Chromosome Location:
          Chr.2: 31867823 - 32011008 on Build GRCh38
          Chromosome Location:
          Chr.21: 36130472 - 36131916 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs04398857_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MEMO1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001137602.4 NP_001131074.1 7 - 8 1450 99
          NM_001301833.4 NP_001288762.1 9 - 10 842 99
          NM_001301852.4 NP_001288781.1 7 - 8 621 99
          NM_001371912.2 NP_001358841.1 9 - 10 1639 99
          NM_001371913.2 NP_001358842.1 9 - 10 971 99
          NM_001371914.2 NP_001358843.1 8 - 9 790 99
          NM_001371916.2 NP_001358845.1 7 - 8 721 99
          NM_001371917.2 NP_001358846.1 7 - 8 1376 99
          NM_001371918.2 NP_001358847.1 10 - 11 971 99
          NM_001371920.2 NP_001358849.1 5 - 6 1225 99
          NM_001371921.2 NP_001358850.1 7 - 8 1373 99
          NM_001385196.1 NP_001372125.1 9 - 10 1074 99
          NM_015955.5 NP_057039.1 8 - 9 1519 99
          NR_126032.3 6 - 7 1299 99
          NR_126034.4 6 - 7 544 99
          NR_163996.2 7 - 8 622 99
          NR_163997.2 7 - 8 1442 99
          NR_163998.2 8 - 9 765 99
          XM_011532896.3 XP_011531198.1 6 - 7 590 99
          XM_047444635.1 XP_047300591.1 8 - 9 773 99
          XM_047444636.1 XP_047300592.1 6 - 7 569 99
          GenBank mRNA AB041018.1 - 8 - 9 1175 99
          AB041019.1 - 8 - 9 824 99
          AF132961.1 - 8 - 9 873 99
          AF363446.1 - 8 - 9 1092 99
          AF529368.1 - 8 - 9 758 99
          AK057760.1 - 6 - 7 577 99
          AK290753.1 - 9 - 10 842 99
          AK297128.1 - 7 - 8 817 99
          AL161966.1 - 2 147 99
          BC018733.1 - 8 - 9 1140 99
          BC070046.1 - 9 - 10 915 99
          BC094681.1 - 8 - 9 779 99
          CD250945.1 - 6 - 7 652 99
          Gene Symbol: MEMO1P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA KX533495.1 - 2 774 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MEMO1
          Click here to find siRNAs for Gene MEMO1P1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of microtubule-based process
          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline