| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009431.hg.1 |
ENST00000524175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009431.hg.1 |
ENST00000524175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009431.hg.1 |
ENST00000524175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009431.hg.1 |
ENST00000524175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009432.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166148.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166148.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166148.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166148.hg.1 |
ENST00000330910.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166148.hg.1 |
ENST00000523047.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166148.hg.1 |
NM_001289970.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166148.hg.1 |
TC0500009432.hg.1 |
Clariom D TC |