Search
Search

PABPN1L| Gene Symbol: PABPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080487.2 | NP_001073956.2 | 2 - 3 | 445 | 107 | |
| NM_001294328.1 | NP_001281257.1 | 2 - 3 | 445 | 107 | ||
| XM_017023230.1 | XP_016878719.1 | 4 - 5 | 1700 | 107 | ||
| GenBank mRNA | BC130001.1 | - | 2 - 3 | 445 | 40133435 | 107 |
| BC130002.1 | - | 2 - 3 | 445 | 107 | ||
| EU190484.1 | - | 2 - 3 | 403 | 107 | ||
Set Membership: |
|