| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012937.hg.1 |
ENST00000484779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010557.hg.1 |
ENST00000438986.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012187.hg.1 |
ENST00000495602.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010517.hg.1 |
ENST00000614215.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010517.hg.1 |
NM_001282301.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010517.hg.1 |
NM_001282301.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007940.hg.1 |
ENST00000618526.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180893.hg.1 |
ENST00000614215.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180893.hg.1 |
NM_001282301.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180893.hg.1 |
ENST00000614215.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180893.hg.1 |
NM_001282301.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180893.hg.1 |
TC1600010517.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600180894.hg.1 |
NM_001282301.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180894.hg.1 |
NM_001282301.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180894.hg.1 |
TC1600010517.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900007940.hg.1 |
ENST00000618526.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010557.hg.1 |
ENST00000438986.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012187.hg.1 |
ENST00000495602.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012937.hg.1 |
ENST00000484779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010517.hg.1 |
ENST00000614215.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010517.hg.1 |
NM_001282301.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010517.hg.1 |
NM_001282301.2 |
Refseq |