Search
Search
| Gene Symbol: MT1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301267.1 | NP_001288196.1 | 3 | 317 | 101 | |
| NM_005950.2 | NP_005941.1 | 3 | 314 | 101 | ||
| GenBank mRNA | BC020757.1 | - | 3 | 317 | 4767886 | 101 |
| BC048334.1 | - | 1 | 1230 | 101 | ||
| BM817643.1 | - | 3 | 317 | 101 | ||
| CD175975.1 | - | 3 | 333 | 101 | ||
Set Membership: |
|