| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168405.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168405.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168405.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168405.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168405.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168405.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168405.hg.1 |
TC1600009040.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600168408.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168408.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168408.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168408.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168408.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168408.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168408.hg.1 |
TC1600009040.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600168413.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168413.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168413.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168413.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168413.hg.1 |
TC1600009040.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009040.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168410.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168410.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168410.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168410.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168410.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168410.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168410.hg.1 |
TC1600009040.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600168404.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168404.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168404.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168404.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168404.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168404.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168404.hg.1 |
TC1600009040.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600168407.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168407.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168407.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168407.hg.1 |
ENST00000442039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168407.hg.1 |
ENST00000563974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600168407.hg.1 |
NM_001272051.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600168407.hg.1 |
TC1600009040.hg.1 |
Clariom D TC |