Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04405012_g1
          See other PTOV1-AS2 GE Assays ›
          Gene Symbol:
          PTOV1-AS2
          Gene Name:
          PTOV1 antisense RNA 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 49856465 - 49859289 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04405012_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTOV1-AS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110730.1 2 - 3 337 89
          GenBank mRNA CK903988.1 - 2 - 3 337 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY051614.1 - 1 - 2 342 Chr. 19 - 49858553 - 49859186 [-] on Build GRCh38 89
          GenBankmRNA BM021429.1 - 2 - 3 321 Chr. 19 - 49856987 - 49859273 [-] on Build GRCh38 89
          GenBankmRNA AW503488.1 - 1 - 2 125 Chr. 19 - 49858212 - 49858964 [-] on Build GRCh38 89
          GenBankmRNA CN271573.1 - 1 - 2 182 Chr. 19 - 49858295 - 49859026 [-] on Build GRCh38 89
          GenBankmRNA BM053079.1 - 2 - 3 321 Chr. 19 - 49858344 - 49859273 [-] on Build GRCh38 89
          GenBankmRNA BM023494.1 - 2 - 3 337 Chr. 19 - 49858344 - 49859289 [-] on Build GRCh38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 4749 MIR4749 mir-4749 -3.9kb Anti-sense Chr. 19 - 49854591 - 49854651 on NCBI Build GRCh38
          Downstream polynucleotide kinase 3'-phosphatase PNKP AOA4,
          EIEE10,
          MCSZ,
          PNK
          2.1kb Sense Chr. 19 - 49861203 - 49867565 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900012004.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 ENST00000601893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Clariom D Assay TC1900012004.hg.1 NR_110730.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900207165.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900207165.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900207165.hg.1 NR_110730.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900207165.hg.1 TC1900012004.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900207165.hg.1 ENST00000593654.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900207165.hg.1 ENST00000599259.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900207165.hg.1 NR_110730.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline