| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007263.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007263.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007263.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007263.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007263.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007263.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007263.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007263.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007263.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800149898.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800149898.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800149898.hg.1 |
TC0800007263.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800149899.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800149899.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800149899.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800149899.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800149899.hg.1 |
TC0800007263.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800149903.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800149903.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800149903.hg.1 |
ENST00000519189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800149903.hg.1 |
NR_125746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800149903.hg.1 |
TC0800007263.hg.1 |
Clariom D TC |