| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006552.hg.1 |
ENST00000628509.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006552.hg.1 |
ENST00000628509.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006552.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006552.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006552.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006552.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144708.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144708.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144708.hg.1 |
TC0600006552.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600006552.hg.1 |
ENST00000628509.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006552.hg.1 |
ENST00000628509.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006552.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006552.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006552.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006552.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144705.hg.1 |
ENST00000628509.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600144705.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144705.hg.1 |
ENST00000628509.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600144705.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144705.hg.1 |
TC0600006552.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600144706.hg.1 |
ENST00000628509.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600144706.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144706.hg.1 |
ENST00000628509.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600144706.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144706.hg.1 |
TC0600006552.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600144707.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144707.hg.1 |
NR_125804.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600144707.hg.1 |
TC0600006552.hg.1 |
Clariom D TC |