Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04935481_m1
          See other GRAMD2B GE Assays ›
          Gene Symbol:
          GRAMD2B
          Gene Name:
          GRAM domain containing 2B
          Gene Aliases:
          GRAMD3, NS3TP2
          Chromosome Location:
          Chr.5: 126360120 - 126494364 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04935481_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GRAMD2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146319.3 NP_001139791.1 6 - 7 836 71
          NM_001146320.3 NP_001139792.1 5 - 6 665 71
          NM_001146321.3 NP_001139793.1 7 - 8 820 71
          NM_001146322.3 NP_001139794.1 6 - 7 616 71
          NM_001349541.2 NP_001336470.1 6 - 7 777 71
          NM_001349542.2 NP_001336471.1 6 - 7 826 71
          NM_001349543.2 NP_001336472.1 5 - 6 665 71
          NM_001349544.2 NP_001336473.1 7 - 8 820 71
          NM_023927.4 NP_076416.2 6 - 7 777 71
          NR_146195.2 7 - 8 820 71
          NR_146196.2 6 - 7 777 71
          NR_146197.2 5 - 6 665 71
          XM_005272057.5 XP_005272114.1 6 - 7 751 71
          XM_005272058.5 XP_005272115.1 6 - 7 751 71
          XM_011543591.3 XP_011541893.2 7 - 8 610 71
          XM_011543592.4 XP_011541894.2 7 - 8 610 71
          XM_011543593.3 XP_011541895.1 6 - 7 836 71
          XM_011543594.3 XP_011541896.1 6 - 7 616 71
          XM_017009781.2 XP_016865270.1 7 - 8 879 71
          XM_047417608.1 XP_047273564.1 6 - 7 836 71
          XM_047417609.1 XP_047273565.1 7 - 8 820 71
          XM_047417610.1 XP_047273566.1 6 - 7 777 71
          XM_047417611.1 XP_047273567.1 6 - 7 13088 71
          XM_047417612.1 XP_047273568.1 6 - 7 13088 71
          XM_047417613.1 XP_047273569.1 7 - 8 13082 71
          XM_047417614.1 XP_047273570.1 7 - 8 659 71
          GenBank mRNA AK024966.1 - 6 - 7 777 71
          AK225829.1 - 6 - 7 777 71
          AK293307.1 - 5 - 6 602 71
          AK294656.1 - 6 - 7 659 71
          AK296267.1 - 7 - 8 757 71
          AK298009.1 - 6 - 7 594 71
          AK298349.1 - 6 - 7 682 71
          AK300154.1 - 6 - 7 805 71
          AK303857.1 - 6 - 7 611 71
          AK316288.1 - 6 - 7 824 71
          AY116970.1 - 6 - 7 579 71
          BC008590.1 - 6 - 7 742 71

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GRAMD2B

          Gene Ontology Categories:

          Process(es)

          protein binding
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline