| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
ENST00000535837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
ENST00000417443.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008512.hg.1 |
NR_024359.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000433425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
ENST00000535837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010889.hg.1 |
NR_024493.2 |
Refseq |