| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010556.hg.1 |
ENST00000580501.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010556.hg.1 |
ENST00000580501.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010556.hg.1 |
ENST00000580501.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000628708.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000628708.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000628708.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000629029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000629029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000629029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007523.hg.1 |
ENST00000629029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011188.hg.1 |
ENST00000458273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
ENST00000413075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007823.hg.1 |
NR_033405.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010639.hg.1 |
ENST00000454614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000626319.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000628708.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000628708.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000628708.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000629029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000629029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000629029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007523.hg.1 |
ENST00000629029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010556.hg.1 |
ENST00000580501.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010556.hg.1 |
ENST00000580501.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010556.hg.1 |
ENST00000580501.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010556.hg.1 |
NR_033407.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011338.hg.1 |
ENST00000457530.1 |
Ensembl |