Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04961799_s1
          See other B3GALT1 GE Assays ›
          Gene Symbol:
          B3GALT1
          Gene Name:
          beta-1,3-galactosyltransferase 1
          Gene Aliases:
          beta3Gal-T1
          Chromosome Location:
          Chr.2: 167293060 - 167870856 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04961799_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B3GALT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020981.3 NP_066191.1 2 1023 125
          XM_005246931.3 XP_005246988.1 5 1457 125
          XM_006712819.3 XP_006712882.1 3 5731 125
          XM_011512085.2 XP_011510387.1 5 1473 125
          GenBank mRNA AF117222.1 - 1 764 125
          BC101545.1 - 1 723 8069051 125
          BC104813.1 - 1 723 8143816 125
          E07739.1 - 5 1347 125

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B3GALT1

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          oligosaccharide biosynthetic process
          UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000430546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000430546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000430546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000430546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 ENST00000392690.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Clariom D Assay TC0200009889.hg.1 NM_020981.3 Refseq
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000430546.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000430546.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000430546.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000430546.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 ENST00000436982.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 NR_131227.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014831.hg.1 NR_131227.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline