Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04963121_s1
          See other ZNF280B GE Assays ›
          Gene Symbol:
          ZNF280B
          Gene Name:
          zinc finger protein 280B
          Gene Aliases:
          5'OY11.1, D87009.C22.3, SUHW2, ZNF279, ZNF632
          Chromosome Location:
          Chr.22: 120078 - 144822 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04963121_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZNF280B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080764.4 NP_542942.2 4 556 99
          NR_130642.2 4 556 99
          XM_011529897.3 XP_011528199.1 3 991 99
          XM_047441143.1 XP_047297099.1 3 495 99
          XM_047441144.1 XP_047297100.1 3 627 99
          XM_047441145.1 XP_047297101.1 5 628 99
          XM_047441146.1 XP_047297102.1 4 688 99
          GenBank mRNA AK097608.1 - 4 1001 99
          BC040020.2 - 4 547 99
          CR456614.1 - 1 277 99

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ZNF280B

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000613655.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000613655.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000613655.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000613655.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000613655.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000613655.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000613655.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000613655.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000619852.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 ENST00000626650.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NM_080764.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130642.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130643.1 Refseq
          Affymetrix Human Clariom D Assay TC2200008134.hg.1 NR_130643.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline