| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000564218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000564218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000564218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000564218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000315678.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000564218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000564218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000564218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000564218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
ENST00000618063.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010023.hg.1 |
NM_001136509.2 |
Refseq |