| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000412944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000423168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000428590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000433396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
ENST00000435389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007856.hg.1 |
NR_110156.1 |
Refseq |