| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000444524.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000444524.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000444524.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000444524.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000491996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000491996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000491996.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000316997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000409864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000444524.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000444524.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000444524.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000444524.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000480583.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000491996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000491996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000491996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
ENST00000611254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016604.hg.1 |
NM_001080504.2 |
Refseq |