| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000423778.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000503669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000506319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
ENST00000508956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012504.hg.1 |
NM_178554.5 |
Refseq |