| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000445998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000445998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000445998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
ENST00000445998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000311659.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000445998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000445998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000445998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
ENST00000445998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009931.hg.1 |
NM_017481.3 |
Refseq |