| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000506739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000506739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000506739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000502759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000506739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000506739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000506739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000511200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
ENST00000512754.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006920.hg.1 |
NR_033931.1 |
Refseq |