| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000514851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000514851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000514851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000514851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000379445.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000395322.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000514851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000514851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000514851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000514851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
ENST00000620385.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001048252.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008560.hg.1 |
NM_001127385.1 |
Refseq |