| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000426471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000435629.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
ENST00000606476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012444.hg.1 |
NR_104061.1 |
Refseq |