Search
Search

ZNF37A| Gene Symbol: ZNF37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007094.4 | NP_001007095.1 | 8 | 8037 | 101 | |
| NM_001178101.3 | NP_001171572.1 | 7 | 7559 | 101 | ||
| NM_001324245.3 | NP_001311174.1 | 7 | 7488 | 101 | ||
| NM_001324246.3 | NP_001311175.1 | 8 | 8033 | 101 | ||
| NM_001324247.3 | NP_001311176.1 | 8 | 7778 | 101 | ||
| NM_001324248.3 | NP_001311177.1 | 7 | 7743 | 101 | ||
| NM_001324249.3 | NP_001311178.1 | 7 | 7739 | 101 | ||
| NM_001324250.3 | NP_001311179.1 | 8 | 7853 | 101 | ||
| NM_001324251.3 | NP_001311180.1 | 8 | 7857 | 101 | ||
| NM_001324259.3 | NP_001311188.1 | 8 | 8087 | 101 | ||
| NM_001324260.3 | NP_001311189.1 | 7 | 7609 | 101 | ||
| NM_003421.4 | NP_003412.1 | 7 | 7563 | 101 | ||
| XM_005252586.5 | XP_005252643.1 | 2 | 7166 | 101 | ||
| XM_011519656.3 | XP_011517958.1 | 8 | 7958 | 101 | ||
| XM_011519657.3 | XP_011517959.1 | 8 | 7782 | 101 | ||
| XM_011519658.4 | XP_011517960.1 | 7 | 7484 | 101 | ||
| XM_047425722.1 | XP_047281678.1 | 8 | 7962 | 101 | ||
Set Membership: |
|