| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |