Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04984709_s1
          See other PRSS35 GE Assays ›
          Gene Symbol:
          PRSS35
          Gene Name:
          protease, serine 35
          Gene Aliases:
          C6orf158, dJ223E3.1
          Chromosome Location:
          Chr.6: 83512475 - 83525704 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04984709_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRSS35
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170423.1 NP_001163894.1 3 1465 85
          NM_153362.2 NP_699193.2 2 1360 85
          GenBank mRNA AK291928.1 - 2 1245 85
          AY358661.1 - 3 1394 85
          BC037170.1 - 2 1283 5241063 85

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PRSS35

          Panther Classification:

          Molecular Function -

          serine protease

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 ENST00000369700.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 ENST00000369700.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 ENST00000369700.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 NM_153362.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 NM_153362.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 NM_153362.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008670.hg.1 NM_153362.2 Refseq
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 ENST00000369700.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 ENST00000369700.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 ENST00000369700.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 NM_001170423.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 NM_153362.2 Refseq
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 NM_153362.2 Refseq
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 NM_153362.2 Refseq
          Affymetrix Human Clariom D Assay TC0600008670.hg.1 NM_153362.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline