| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000451294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000451294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000451294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000451294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000487744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000487744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000487744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
ENST00000487744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000274990.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000372569.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000416431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000451294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000451294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000451294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000451294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000477866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000485819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000487744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000487744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000487744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
ENST00000487744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014293.hg.1 |
NM_206922.2 |
Refseq |