Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04994163_m1
          See other GTF2E2 GE Assays ›
          Gene Symbol:
          GTF2E2
          Gene Name:
          general transcription factor IIE subunit 2
          Gene Aliases:
          FE, TF2E2, TFIIE-B, TTD6
          Chromosome Location:
          Chr.8: 30578318 - 30658236 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04994163_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GTF2E2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001348353.1 NP_001335282.1 5 - 6 599 120
          NM_002095.6 NP_002086.1 5 - 6 819 120
          XM_011544509.3 XP_011542811.1 5 - 6 819 120
          XM_011544510.2 XP_011542812.1 5 - 6 819 120
          XM_017013363.2 XP_016868852.1 5 - 6 664 120
          XM_017013364.2 XP_016868853.1 5 - 6 599 120
          XM_024447138.2 XP_024302906.1 5 - 6 834 120
          GenBank mRNA BC030572.2 - 5 - 6 798 120
          BX421791.2 - 5 - 6 120
          S67861.1 - 5 - 6 741 120
          X63469.1 - 5 - 6 793 120

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GTF2E2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          TFIIH-class transcription factor binding
          DNA binding
          RNA binding
          protein binding
          obsolete general RNA polymerase II transcription factor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518445.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518445.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518445.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518445.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518445.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518445.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518445.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518445.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
          Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline