Search
Search

FAM83A| Gene Symbol: FAM83A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321630.1 | NP_001308559.1 | 2 - 3 | 1150 | 73 | |
| NM_032899.6 | NP_116288.2 | 3 - 4 | 1574 | 73 | ||
| NM_207006.2 | NP_996889.1 | 3 - 4 | 2659 | 73 | ||
| XM_011517338.1 | XP_011515640.1 | 2 - 3 | 947 | 73 | ||
| GenBank mRNA | AF497803.1 | - | 2 - 3 | 899 | 73 | |
| AK095301.1 | - | 3 - 4 | 2659 | 73 | ||
| AK098407.1 | - | 2 - 3 | 248 | 73 | ||
| BC052300.1 | - | 2 - 3 | 963 | 6421272 | 73 | |
| DQ280323.1 | - | 3 - 4 | 1547 | 73 | ||
Set Membership: |
|