| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000527445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000527445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000527445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000527445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000284486.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000527445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000527445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000527445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000527445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528111.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000528897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000531804.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
ENST00000534308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009600.hg.1 |
NM_053279.2 |
Refseq |