Search
Search

LOC107984125| Gene Symbol: LOC107984125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017014157.1 | XP_016869646.1 | 23 - 24 | 3292 | 121 | |
| XM_017014158.1 | XP_016869647.1 | 18 - 19 | 2628 | 121 | ||
Set Membership: |
|