| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000455039.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000455039.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000455039.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000455039.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000603893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000605260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000605260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
ENST00000605260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000455039.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000455039.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000455039.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000455039.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000603893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000605260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000605260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
ENST00000605260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011936.hg.1 |
NR_038969.1 |
Refseq |